Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013103.2 | NP_001013121.2 | 4, 5 | 508 | ||
GenBank mRNA | CB611861.1 | - | 4, 5 | 511 | ||
FQ228127.1 | - | 1, 2 | 37 |