Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318339.1 | NP_001305268.1 | 16 | 3057 | ||
NM_015308.3 | NP_056123.2 | 16 | 3051 | |||
GenBank mRNA | AB023231.2 | - | 16 | 3050 | ||
AK022987.1 | - | 9 | 1495 | |||
AL137480.1 | - | 16 | 2850 | |||
BC037404.1 | - | 16 | 2882 | 5140963 |