Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pyy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034080.1 | NP_001029252.1 | 2, 3 | 263 | ||
GenBank mRNA | AB238226.1 | - | 2, 3 | 263 | ||
BF551062.1 | - | 2, 3 | 270 | |||
M17523.1 | - | 1, 2 | 242 |