Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128155.1 | NP_001121627.1 | 2 | 438 | ||
GenBank mRNA | EF532356.1 | - | 2 | 328 | ||
EF532357.1 | - | 2 | 320 | |||
EF532358.1 | - | 2 | 320 | |||
EF532359.1 | - | 2 | 328 | |||
EF532360.1 | - | 2 | 320 |