Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126081.1 | NP_001119553.1 | 3 | 116 | ||
GenBank mRNA | AY310147.1 | - | 3 | 116 |