Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134698.1 | NP_001128170.1 | 15 | 2957 | ||
GenBank mRNA | BC168153.1 | - | 15 | 2957 |