Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053873.1 | NP_446325.1 | 3660 | |||
GenBank mRNA | AB374360.1 | - | 3660 | |||
L04684.1 | - | 1354 | ||||
U31816.1 | - | 3 | 281 |