Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135896.1 | NP_001129368.1 | 5 | 729 | ||
XM_008765581.1 | XP_008763803.1 | 3 | 547 | |||
XM_008765582.1 | XP_008763804.1 | 3 | 566 | |||
XM_008765583.1 | XP_008763805.1 | 4 | 1270 | |||
GenBank mRNA | BC127521.1 | - | 5 | 729 | 7110443 |