Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vpreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108845.1 | NP_001102315.1 | 2 | 261 |
Gene Symbol: Vpreb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134788.1 | NP_001128260.1 | 2 | 137 |