Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly49si2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009498.1 | NP_001009498.1 | 457 | |||
XM_008763406.1 | XP_008761628.1 | 470 | ||||
XM_008763407.1 | XP_008761629.1 | 470 | ||||
XM_008763408.1 | XP_008761630.1 | 470 | ||||
XM_008763409.1 | XP_008761631.1 | 470 | ||||
GenBank mRNA | AY659933.1 | - | 457 |
Gene Symbol: Klra17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172088.1 | NP_001165559.1 | 4 | 460 |
Gene Symbol: Klra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009718.1 | NP_001009718.1 | 5 | 581 | ||
XM_008763403.1 | XP_008761625.1 | 4 | 643 | |||
GenBank mRNA | AY651018.1 | - | 5 | 581 |
Gene Symbol: Klra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198746.1 | NP_942041.1 | 5 | 597 | ||
XM_008763341.1 | XP_008761563.1 | 5 | 753 | |||
XM_008763342.1 | XP_008761564.1 | 5 | 573 | |||
XM_008763343.1 | XP_008761565.1 | 5 | 483 | |||
XM_008763344.1 | XP_008761566.1 | 784 | ||||
GenBank mRNA | U56863.1 | - | 5 | 597 |
Gene Symbol: LOC100910264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763346.1 | XP_008761568.1 | 4 | 3036 |
Gene Symbol: LOC497796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111310.1 | NP_001104780.1 | 616 | |||
XM_006237126.2 | XP_006237188.1 | 5 | 666 | |||
GenBank mRNA | BC093399.1 | - | 616 | 7389533 |
Gene Symbol: LOC502907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763434.1 | XP_008761656.1 | 5 | 1139 |
Gene Symbol: LOC502908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763439.1 | XP_008761661.1 | 3 | 794 | ||
GenBank mRNA | AY669088.1 | - | 3 | 332 |
Gene Symbol: LOC690097 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763433.1 | XP_008761655.1 | 5 | 883 |
Gene Symbol: Ly49i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009499.1 | NP_001009499.1 | 457 | |||
XM_008763436.1 | XP_008761658.1 | 3 | 660 | |||
XM_008763437.1 | XP_008761659.1 | 2 | 766 | |||
XM_008763438.1 | XP_008761660.1 | 3 | 724 | |||
GenBank mRNA | AY659932.1 | - | 457 |
Gene Symbol: Ly49i4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009495.1 | NP_001009495.1 | 4 | 544 | ||
XM_008763405.1 | XP_008761627.1 | 5 | 659 | |||
GenBank mRNA | AY649834.1 | - | 4 | 544 |
Gene Symbol: Ly49i9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009496.1 | NP_001009496.1 | 5 | 623 | ||
GenBank mRNA | AY649839.1 | - | 5 | 623 |
Gene Symbol: Ly49s3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153726.1 | NP_714948.1 | 6 | 585 | ||
GenBank mRNA | AY102036.1 | - | 6 | 585 | ||
AY747628.1 | - | 7 | 636 |
Gene Symbol: Ly49s4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009487.1 | NP_001009487.1 | 585 | |||
GenBank mRNA | AY649831.1 | - | 585 |
Gene Symbol: Ly49s7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009494.1 | NP_001009494.1 | 5 | 595 | ||
GenBank mRNA | AY649833.1 | - | 5 | 595 | ||
AY649840.1 | - | 5 | 587 |
Gene Symbol: Ly49si3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009919.1 | NP_001009919.1 | 4 | 457 | ||
XM_006237116.2 | XP_006237178.1 | 551 | ||||
XM_008763410.1 | XP_008761632.1 | 5 | 738 | |||
XM_008763411.1 | XP_008761633.1 | 5 | 1375 | |||
XM_008763412.1 | XP_008761634.1 | 5 | 532 | |||
XM_008763413.1 | XP_008761635.1 | 5 | 738 | |||
XM_008763414.1 | XP_008761636.1 | 5 | 738 | |||
XM_008763415.1 | XP_008761637.1 | 5 | 738 | |||
GenBank mRNA | AY653730.1 | - | 4 | 457 |