Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr25l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135894.1 | NP_001129366.1 | 1 | 79 | ||
XM_006240544.2 | XP_006240606.1 | 2 | 296 | |||
XM_006240545.2 | XP_006240607.1 | 2 | 141 | |||
XM_006240546.2 | XP_006240608.1 | 2 | 159 | |||
XM_006240547.2 | XP_006240609.1 | 2 | 367 | |||
GenBank mRNA | BC166712.1 | - | 1 | 79 | 7930656 |