Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015329.3 | NP_056144.3 | 3 | 345 | ||
XM_005259837.3 | XP_005259894.2 | 3 | 337 | |||
XM_006722711.3 | XP_006722774.2 | 3 | 337 | |||
XM_011527844.2 | XP_011526146.1 | 3 | 750 | |||
GenBank mRNA | BY795974.2 | - | 3 | 313 |