Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATP13A2 SIRNA Assays ›
Gene Symbol:
ATP13A2
Gene Name:
ATPase 13A2
Gene Aliases:
CLN12, HSA9947, KRPPD, PARK9
Chromosome Location:
Chr.1: 16985958 - 17011972 on Build GRCh38
UniGene ID:
Hs.128866
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23744
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATP13A2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001141973.2
NP_001135445.1
14, 15
1583
NM_001141974.2
NP_001135446.1
14, 15
1583
NM_022089.3
NP_071372.1
14, 15
1598
XM_005245810.1
XP_005245867.1
14, 15
1595
XM_005245811.1
XP_005245868.1
14, 15
1583
XM_005245812.1
XP_005245869.1
14, 15
1571
XM_005245815.1
XP_005245872.1
14, 15
1598
XM_006710512.1
XP_006710575.1
14, 15
1580
XM_006710513.1
XP_006710576.1
14, 15
1556
XM_011541128.1
XP_011539430.1
14, 15
1598
XM_011541129.1
XP_011539431.1
14, 15
1598
XM_017000844.1
XP_016856333.1
14, 15
1598
XM_017000845.1
XP_016856334.1
14, 15
1580
XM_017000846.1
XP_016856335.1
14, 15
1556
XM_017000847.1
XP_016856336.1
14, 15
1568
XM_017000848.1
XP_016856337.1
14, 15
1598
XM_017000849.1
XP_016856338.1
14, 15
1583
XM_017000850.1
XP_016856339.1
14, 15
1598
GenBank mRNA
AK075310.1
-
4, 5
380
AK290210.1
-
14, 15
1539
AL354615.1
-
14, 15
1398
AY461712.1
-
14, 15
1397
AY987009.1
-
4, 5
392
BC034575.1
-
11, 12
1089
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cellular calcium ion homeostasis
cellular iron ion homeostasis
cellular zinc ion homeostasis
regulation of mitochondrion organization
regulation of macroautophagy
regulation of autophagosome size
cellular cation homeostasis
regulation of intracellular protein transport
ion transmembrane transport
cellular response to oxidative stress
protein autophosphorylation
positive regulation of protein secretion
regulation of endopeptidase activity
zinc ion homeostasis
cellular response to manganese ion
cellular response to zinc ion
cation transmembrane transport
negative regulation of neuron death
regulation of mitophagy
positive regulation of exosomal secretion
regulation of chaperone-mediated autophagy
autophagosome organization
polyamine import
regulation of lysosomal protein catabolic process
negative regulation of lysosomal protein catabolic process
peptidyl-aspartic acid autophosphorylation
protein binding
ATP binding
zinc ion binding
ATPase activity
cation-transporting ATPase activity
manganese ion binding
phosphatidic acid binding
phosphatidylinositol-3,5-bisphosphate binding
cupric ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0