Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012071.3 | NP_036203.1 | 7, 8 | 555 | ||
GenBank mRNA | AF078848.1 | - | 7, 8 | 530 | ||
AF201948.1 | - | 7, 8 | 549 | |||
AK128789.1 | - | 4, 5 | 1070 | |||
AY542159.1 | - | 7, 8 | 520 | |||
BC022898.2 | - | 7, 8 | 542 | 3531636 | ||
BG496810.1 | - | 7, 8 | 548 |
Gene Symbol: COMMD3-BMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204062.1 | NP_001190991.1 | 5, 6 | 437 | ||
GenBank mRNA | AK313235.1 | - | 5, 6 | 412 |