Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000070.2 | NP_000061.1 | 21 | 2542 | ||
NM_024344.1 | NP_077320.1 | 20 | 2524 | |||
NM_173087.1 | NP_775110.1 | 18 | 2266 | |||
NM_173088.1 | NP_775111.1 | 10 | 869 | |||
NM_173089.1 | NP_775112.1 | 6 | 499 | |||
NM_173090.1 | NP_775113.1 | 7 | 517 | |||
GenBank mRNA | AF127764.1 | - | 20 | 2439 | ||
AF127765.3 | - | 18 | 2181 | |||
AK027249.1 | - | 5 | 1159 | |||
BC003169.1 | - | 10 | 869 | |||
BC003521.1 | - | 6 | 481 | |||
BC004883.2 | - | 6 | 499 | |||
BC007810.2 | - | 7 | 465 | |||
BC067126.1 | - | 6 | 482 | |||
BC100782.1 | - | 6 | 480 | |||
BC100783.1 | - | 6 | 480 | |||
BC100784.2 | - | 6 | 480 | |||
BC107791.1 | - | 7 | 632 | |||
BC128605.1 | - | 6 | 480 | |||
BC146649.1 | - | 21 | 2455 | 40147158 | ||
BC146672.1 | - | 21 | 2455 | 40148444 | ||
BT007322.1 | - | 9 | 700 | |||
EU791850.1 | - | 21 | 1975 | |||
EU791851.1 | - | 20 | 1831 | |||
KU177954.1 | - | 9 | 700 | |||
KU177955.1 | - | 9 | 801 | |||
KU177956.1 | - | 7 | 568 | |||
X85030.1 | - | 21 | 2236 |