Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271684.1 | NP_001258613.1 | 3, 4 | 732 | ||
NM_001271685.1 | NP_001258614.1 | 3, 4 | 567 | |||
NM_012243.2 | NP_036375.1 | 3, 4 | 732 | |||
XM_005270691.4 | XP_005270748.1 | 4, 5 | 1253 | |||
XM_011541136.2 | XP_011539438.1 | 4, 5 | 649 | |||
XM_011541137.2 | XP_011539439.1 | 3, 4 | 709 | |||
XM_011541138.2 | XP_011539440.1 | 3, 4 | 718 | |||
XM_017000870.1 | XP_016856359.1 | 3, 4 | 1154 | |||
GenBank mRNA | AB021981.1 | - | 3, 4 | 537 | ||
AK024989.1 | - | 2, 3 | 297 | |||
AK290573.1 | - | 3, 4 | 490 | |||
BC005136.2 | - | 3, 4 | 515 | 3505772 | ||
CR749816.1 | - | 3, 4 | 567 |