Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012405.3 | NP_036537.1 | 3 | 500 | ||
XM_011541140.2 | XP_011539442.1 | 2 | 200 | |||
GenBank mRNA | AF064084.1 | - | 3 | 470 | ||
AK092890.1 | - | 4 | 441 | |||
BC017037.1 | - | 4 | 595 | |||
BC028168.1 | - | 3 | 456 | 5245125 | ||
CR541671.1 | - | 3 | 427 | |||
CR541711.1 | - | 3 | 427 |