Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303494.1 | NP_001290423.1 | 5 | 1195 | ||
NM_014292.4 | NP_055107.3 | 5 | 1249 | |||
GenBank mRNA | AK293035.1 | - | 5 | 1131 | ||
BC012111.1 | - | 5 | 1128 | 4554483 | ||
BC064900.1 | - | 5 | 1116 | 6009294 |