Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015356.4 | NP_056171.3 | 10 | 940 | ||
NM_182706.4 | NP_874365.3 | 10 | 940 | |||
GenBank mRNA | AF240677.1 | - | 10 | 933 | ||
AF271734.2 | - | 10 | 911 | |||
AY062238.1 | - | 10 | 942 | |||
D63481.3 | - | 10 | 935 |