Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320445.1 | NP_001307374.1 | 7 | 910 | ||
NM_001320446.1 | NP_001307375.1 | 9 | 1069 | |||
NM_015358.2 | NP_056173.1 | 8 | 983 | |||
XM_011529518.2 | XP_011527820.1 | 8 | 1203 | |||
XM_017028311.1 | XP_016883800.1 | 8 | 1005 | |||
XM_017028312.1 | XP_016883801.1 | 8 | 926 | |||
GenBank mRNA | AK292957.1 | - | 8 | 934 | ||
AK295133.1 | - | 7 | 902 | |||
BC007311.1 | - | 8 | 940 | |||
BC094779.1 | - | 8 | 940 | 30527767 | ||
BC132731.1 | - | 8 | 937 | 40146753 | ||
BC144116.1 | - | 9 | 1023 | |||
D50926.2 | - | 8 | 942 |