Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNPO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191028.2 | NP_001177957.2 | 12 | 1930 | ||
NM_012470.3 | NP_036602.1 | 13 | 2122 | |||
NR_034053.2 | 14 | 2283 | ||||
XM_011515989.2 | XP_011514291.1 | 13 | 1724 | |||
GenBank mRNA | AF145029.1 | - | 13 | 1934 | ||
AJ133769.1 | - | 13 | 1719 | |||
AK001263.1 | - | 6 | 676 | |||
AK022910.1 | - | 4 | 456 | |||
AK225999.1 | - | 12 | 1901 | |||
AK312580.1 | - | 13 | 1849 | |||
BC068530.1 | - | 13 | 1850 |