Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012437.5 | NP_036569.1 | 1 | 212 | ||
NR_052019.1 | 1 | 212 | ||||
GenBank mRNA | AF086837.1 | - | 1 | 122 | ||
AK024555.1 | - | 1 | 189 | |||
BC000761.2 | - | 1 | 141 | 2821705 | ||
BC004494.2 | - | 1 | 142 | 3830132 | ||
BI117540.1 | - | 213 | ||||
BM051828.1 | - | 1 | 198 | |||
BT006753.1 | - | 1 | 122 | |||
CD671119.1 | - | 1 | 147 |