Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012106.3 | NP_036238.1 | 2, 3 | 349 | ||
XM_011522977.1 | XP_011521279.1 | 2, 3 | 88 | |||
GenBank mRNA | AF126062.1 | - | 2, 3 | 208 | ||
AK075050.1 | - | 2, 3 | 349 | |||
AK298758.1 | - | 2, 3 | 99 | |||
AK311018.1 | - | 2, 3 | 241 | |||
BC003087.1 | - | 2, 3 | 221 | 3503251 |