Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARHSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042476.2 | NP_001035941.1 | 4 | 700 | ||
NM_001278260.1 | NP_001265189.1 | 5 | 508 | |||
NM_001278261.1 | NP_001265190.1 | 5 | 647 | |||
NM_001278262.1 | NP_001265191.1 | 5 | 606 | |||
NM_001278263.1 | NP_001265192.1 | 4 | 382 | |||
NM_001278264.1 | NP_001265193.1 | 4 | 524 | |||
NM_001278265.1 | NP_001265194.1 | 4 | 516 | |||
NM_001278266.1 | NP_001265195.1 | 4 | 492 | |||
NM_014316.3 | NP_055131.2 | 4 | 394 | |||
XM_005255228.4 | XP_005255285.3 | 4 | 558 | |||
XM_005255229.4 | XP_005255286.1 | 4 | 370 | |||
XM_011522444.2 | XP_011520746.1 | 4 | 369 | |||
GenBank mRNA | AF115345.1 | - | 3 | 351 | ||
AK311777.1 | - | 4 | 388 | |||
BC001333.1 | - | 4 | 700 | |||
BC003366.1 | - | 4 | 391 | 2900952 | ||
BC108283.1 | - | 4 | 374 | 4430943 | ||
BM920489.1 | - | 4 | 382 | |||
CA396216.1 | - | 4 | 451 | |||
DB101255.1 | - | 5 | 502 | |||
HY090855.1 | - | 4 | 394 |