Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167614.1 | NP_001161086.1 | 5 | 1745 | ||
NM_014319.4 | NP_055134.2 | 5 | 1748 | |||
GenBank mRNA | AF112299.2 | - | 5 | 1728 | ||
AK024954.1 | - | 5 | 423 | |||
AK312473.1 | - | 5 | 1745 |