Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105203.1 | NP_001098673.1 | 4 | 1751 | ||
NM_001105204.1 | NP_001098674.1 | 4 | 1751 | |||
NM_001105205.1 | NP_001098675.1 | 3 | 773 | |||
NM_001278227.1 | NP_001265156.1 | 3 | 816 | |||
NM_001278228.1 | NP_001265157.1 | 3 | 816 | |||
NM_001278229.1 | NP_001265158.1 | 3 | 816 | |||
NM_001278230.1 | NP_001265159.1 | 2 | 615 | |||
NM_014328.4 | NP_055143.2 | 3 | 816 | |||
NR_103478.1 | 2 | 481 | ||||
XM_006711254.1 | XP_006711317.1 | 5 | 2162 | |||
XM_006711256.1 | XP_006711319.1 | 5 | 2162 | |||
XM_006711257.1 | XP_006711320.1 | 3 | 430 | |||
XM_017000891.1 | XP_016856380.1 | 4 | 2332 | |||
XM_017000892.1 | XP_016856381.1 | 3 | 430 | |||
GenBank mRNA | AB026894.2 | - | 3 | 240 | ||
AK055451.1 | - | 3 | 1011 | |||
AK074485.1 | - | 2 | 369 | |||
AK074982.1 | - | 3 | 235 | |||
AK093777.1 | - | 3 | 269 | |||
AK125378.1 | - | 4 | 1703 | |||
AK298730.1 | - | 3 | 326 | |||
AK307736.1 | - | 3 | 266 | |||
AK314559.1 | - | 3 | 485 | |||
AL080083.1 | - | 2 | 93 | |||
BC001045.2 | - | 3 | 257 | |||
BC004293.1 | - | 2 | 337 | |||
BC028055.1 | - | 2 | 365 | |||
BC052277.1 | - | 3 | 773 | |||
BX402179.2 | - | 3 | 307 | |||
BX640612.1 | - | 2 | 433 | |||
CN305634.1 | - | 2 | 615 | |||
CR749671.1 | - | 2 | 1322 | |||
DA131200.1 | - | 3 | 326 | |||
DA751328.1 | - | 2 | 447 |