Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012113.2 | NP_036245.1 | 3 | 534 | ||
XM_005245059.2 | XP_005245116.1 | 3 | 463 | |||
XM_005245060.2 | XP_005245117.1 | 3 | 463 | |||
XM_006711259.2 | XP_006711322.1 | 3 | 463 | |||
XM_017000894.1 | XP_016856383.1 | 3 | 463 | |||
XM_017000895.1 | XP_016856384.1 | 3 | 463 | |||
GenBank mRNA | AB025904.1 | - | 3 | 457 | ||
AK074765.1 | - | 3 | 503 | |||
AK290609.1 | - | 3 | 373 | |||
AK315469.1 | - | 3 | 466 | |||
AY358689.1 | - | 3 | 418 | |||
BC034412.1 | - | 3 | 388 | 5175251 | ||
BP361793.1 | - | 3 | 534 | |||
BT020054.1 | - | 3 | 148 |