Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130710.1 | NP_001124182.1 | 2 | 65 | ||
NM_001139499.1 | NP_001132971.1 | 2 | 304 | |||
NM_012322.2 | NP_036454.1 | 2 | 128 | |||
NR_024466.1 | 2 | 875 | ||||
GenBank mRNA | AF182291.1 | - | 2 | 89 | ||
AJ238097.1 | - | 2 | 82 | |||
AK024217.1 | - | 2 | 111 | |||
BC005938.1 | - | 2 | 88 | 4132686 | ||
BF667238.1 | - | 2 | 129 | |||
BF700164.1 | - | 2 | 65 | |||
DA736378.1 | - | 2 | 295 |