Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012320.3 | NP_036452.1 | 2, 3 | 373 | ||
XM_005255886.3 | XP_005255943.1 | 2, 3 | 346 | |||
GenBank mRNA | AB017494.1 | - | 2, 3 | 322 | ||
AK001705.1 | - | 2, 3 | 334 | |||
AK074828.1 | - | 2, 3 | 373 | |||
AK296263.1 | - | 2, 3 | 344 | |||
AL110209.1 | - | 2, 3 | 354 | |||
AY358425.1 | - | 2, 3 | 310 | |||
BC062605.1 | - | 2, 3 | 337 | 5162981 |