Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABARAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031412.2 | NP_113600.1 | 3, 4 | 533 | ||
GenBank mRNA | AF087847.1 | - | 3, 4 | 574 | ||
AF287012.1 | - | 3, 4 | 624 | |||
AK056846.1 | - | 5, 6 | 694 | |||
AK074195.1 | - | 3, 4 | 550 | |||
AK092075.1 | - | 3, 4 | 963 | |||
AK315418.1 | - | 3, 4 | 546 | |||
AL136676.1 | - | 3, 4 | 519 | |||
BC009309.2 | - | 3, 4 | 530 | 4100219 | ||
BC028315.1 | - | 3, 4 | 533 | 5183078 | ||
BX648781.1 | - | 4, 5 | 1911 | |||
CR533480.1 | - | 3, 4 | 293 |