Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033054.2 | NP_001028226.1 | 4 | 600 | ||
NM_001033055.2 | NP_001028227.1 | 4 | 609 | |||
NM_001285399.2 | NP_001272328.1 | 5 | 753 | |||
NM_001285400.2 | NP_001272329.1 | 5 | 723 | |||
NM_001285402.1 | NP_001272331.1 | 5 | 780 | |||
NM_001285403.2 | NP_001272332.1 | 5 | 765 | |||
NM_014336.4 | NP_055151.3 | 5 | 789 | |||
GenBank mRNA | AB593042.1 | - | 5 | 686 | ||
AB593043.1 | - | 5 | 710 | |||
AB593044.1 | - | 5 | 724 | |||
AB593045.1 | - | 5 | 724 | |||
AB593046.1 | - | 5 | 726 | |||
AB593047.1 | - | 5 | 767 | |||
AB593048.1 | - | 5 | 780 | |||
AB593049.1 | - | 5 | 734 | |||
AB593050.1 | - | 5 | 710 | |||
AB593051.1 | - | 5 | 711 | |||
AB593052.1 | - | 5 | 710 | |||
AB593053.1 | - | 4 | 566 | |||
AB593054.1 | - | 4 | 509 | |||
AB593055.1 | - | 5 | 755 | |||
AF038437.1 | - | 5 | 708 | |||
AF148864.1 | - | 5 | 874 | |||
AF525970.1 | - | 4 | 600 | |||
AJ633677.1 | - | 4 | 562 | |||
AJ633678.1 | - | 4 | 553 | |||
AJ830742.1 | - | 5 | 677 | |||
AK023970.1 | - | 5 | 717 | |||
AK314558.1 | - | 5 | 710 | |||
BC012055.1 | - | 5 | 702 | 4634156 | ||
BX537907.1 | - | 5 | 687 | |||
BX647279.1 | - | 5 | 755 |