Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317996.1 | NP_001304925.1 | 10 | 698 | ||
NM_014337.3 | NP_055152.1 | 10 | 698 | |||
NM_148175.2 | NP_680480.1 | 10 | 698 | |||
NM_148176.2 | NP_680481.1 | 10 | 698 | |||
XM_005261448.3 | XP_005261505.1 | 10 | 698 | |||
XM_011530041.2 | XP_011528343.1 | 10 | 698 | |||
XM_011530042.2 | XP_011528344.1 | 10 | 698 | |||
XM_011530043.2 | XP_011528345.1 | 10 | 698 | |||
XM_011530044.2 | XP_011528346.1 | 10 | 698 | |||
XM_011530046.2 | XP_011528348.1 | 10 | 698 | |||
XM_011530047.2 | XP_011528349.1 | 10 | 698 | |||
XM_011530049.2 | XP_011528351.1 | 11 | 748 | |||
XM_011530050.2 | XP_011528352.1 | 9 | 611 | |||
XM_011530051.2 | XP_011528353.1 | 10 | 638 | |||
GenBank mRNA | AK127259.1 | - | 11 | 721 | ||
AK289545.1 | - | 10 | 660 | |||
AK299206.1 | - | 9 | 560 | |||
BC000022.2 | - | 10 | 660 | 3505936 | ||
BC028385.2 | - | 10 | 642 | 4816793 | ||
CR456548.1 | - | 10 | 614 | |||
U37219.1 | - | 10 | 674 |