Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OSBP2 SIRNA Assays ›
Gene Symbol:
OSBP2
Gene Name:
oxysterol binding protein 2
Gene Aliases:
HLM, ORP-4, ORP4, OSBPL1, OSBPL4
Chromosome Location:
Chr.22: 30693782 - 30907824 on Build GRCh38
UniGene ID:
Hs.517546
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24369
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OSBP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282738.1
NP_001269667.1
11
1834
NM_001282739.1
NP_001269668.1
10
2154
NM_001282740.1
NP_001269669.1
9
1337
NM_001282741.1
NP_001269670.1
9
1394
NM_001282742.1
NP_001269671.1
8
1065
NM_030758.3
NP_110385.1
10
2157
XM_005261465.4
XP_005261522.1
9
1936
XM_006724203.2
XP_006724266.2
9
1933
XM_006724207.2
XP_006724270.1
9
1340
XM_006724208.3
XP_006724271.1
8
1125
XM_011530057.1
XP_011528359.1
10
2053
XM_011530058.1
XP_011528360.1
10
2050
XM_011530059.2
XP_011528361.1
10
2083
XM_011530060.1
XP_011528362.1
9
1394
XM_011530061.1
XP_011528363.1
8
1038
XM_011530062.1
XP_011528364.1
10
1439
XM_017028708.1
XP_016884197.1
9
1936
XM_017028709.1
XP_016884198.1
10
2080
XM_017028710.1
XP_016884199.1
10
1546
XM_017028711.1
XP_016884200.1
8
1697
XM_017028712.1
XP_016884201.1
9
1341
XM_017028713.1
XP_016884202.1
9
1667
GenBank mRNA
AB051451.2
-
10
2065
AF288741.1
-
10
2051
AF323731.1
-
10
2157
AK093196.1
-
9
1337
AK131374.1
-
10
1684
AK294007.1
-
9
1942
AK296354.1
-
9
1394
AK296525.1
-
11
1836
AK300323.1
-
8
1065
BC004340.1
-
7
952
BC118914.2
-
10
2071
40037933
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
lipid transport
spermatid development
protein binding
cholesterol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0