Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASQ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001232.3 | NP_001223.2 | 1 | 444 | ||
GenBank mRNA | AK129891.1 | - | 1 | 419 | ||
AK225959.1 | - | 1 | 306 | |||
AK293595.1 | - | 1 | 453 | |||
AK295502.1 | - | 1 | 353 | |||
AK313041.1 | - | 1 | 390 | |||
BC022288.1 | - | 1 | 353 | 4289141 | ||
D55655.1 | - | 1 | 289 | |||
DC423064.1 | - | 1 | 444 |