Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167621.1 | NP_001161093.1 | 10 | 1415 | ||
NM_013236.3 | NP_037368.1 | 11 | 1607 | |||
GenBank mRNA | AF086923.1 | - | 11 | 1560 | ||
AF119662.1 | - | 11 | 1547 | |||
AK025734.1 | - | 11 | 1538 | |||
AK294348.1 | - | 10 | 1408 | |||
AL050282.1 | - | 11 | 1539 | |||
AL137374.1 | - | 2 | 176 | |||
BC007508.2 | - | 11 | 1529 | 3030062 | ||
CR456568.1 | - | 11 | 1392 | |||
CR457381.1 | - | 11 | 1341 |