Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015382.3 | NP_056197.3 | 19 | 3419 | ||
XM_005267502.2 | XP_005267559.2 | 19 | 3581 | |||
XM_011536619.2 | XP_011534921.1 | 19 | 3017 | |||
XM_011536621.2 | XP_011534923.1 | 19 | 3017 | |||
XM_011536622.1 | XP_011534924.1 | 10 | 1344 | |||
XM_017021148.1 | XP_016876637.1 | 18 | 2911 | |||
XM_017021149.1 | XP_016876638.1 | 18 | 2911 | |||
XM_017021150.1 | XP_016876639.1 | 18 | 2911 | |||
XM_017021151.1 | XP_016876640.1 | 19 | 3613 | |||
XM_017021152.1 | XP_016876641.1 | 19 | 3613 | |||
XM_017021153.1 | XP_016876642.1 | 19 | 3017 | |||
GenBank mRNA | AB032957.2 | - | 11 | 1565 | ||
AK095634.1 | - | 10 | 1312 | |||
AY254380.1 | - | 19 | 3219 |