Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARMC8 SIRNA Assays ›
Gene Symbol:
ARMC8
Gene Name:
armadillo repeat containing 8
Gene Aliases:
GID5, HSPC056, S863-2, VID28
Chromosome Location:
Chr.3: 138187248 - 138298389 on Build GRCh38
UniGene ID:
Hs.744868
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24624
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARMC8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001267041.2
NP_001253970.1
6
795
NM_001267042.2
NP_001253971.1
7
888
NM_001282342.1
NP_001269271.1
5
718
NM_014154.4
NP_054873.2
8
964
NM_015396.5
NP_056211.2
8
964
NM_213654.3
NP_998819.1
8
1018
XM_006713564.3
XP_006713627.1
7
883
XM_006713565.2
XP_006713628.1
8
924
XM_006713566.3
XP_006713629.1
6
778
XM_006713567.3
XP_006713630.1
7
883
XM_011512632.1
XP_011510934.1
7
831
XM_011512633.1
XP_011510935.1
7
885
XM_017006095.1
XP_016861584.1
8
886
XM_017006096.1
XP_016861585.1
6
778
XM_017006097.1
XP_016861586.1
7
832
XM_017006098.1
XP_016861587.1
6
778
XM_017006099.1
XP_016861588.1
7
869
XM_017006100.1
XP_016861589.1
7
883
XM_017006101.1
XP_016861590.1
7
793
GenBank mRNA
AF109184.1
-
8
717
AF161541.1
-
8
686
AK001721.1
-
6
526
AK289577.1
-
8
660
AK296757.1
-
7
619
AK296837.1
-
6
495
AK299229.1
-
2
156
AK299769.1
-
5
438
AK304558.1
-
6
471
AK315932.1
-
2
161
BC013424.1
-
8
718
3892143
BC032661.1
-
8
828
5530425
BC041699.1
-
8
643
5240423
CX167429.1
-
7
593
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0