Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267041.2 | NP_001253970.1 | 10 | 1169 | ||
NM_001282342.1 | NP_001269271.1 | 9 | 1092 | |||
NM_014154.4 | NP_054873.2 | 12 | 1338 | |||
NM_015396.5 | NP_056211.2 | 12 | 1338 | |||
NM_213654.3 | NP_998819.1 | 12 | 1392 | |||
XM_006713564.3 | XP_006713627.1 | 11 | 1257 | |||
XM_006713565.2 | XP_006713628.1 | 12 | 1298 | |||
XM_006713566.3 | XP_006713629.1 | 10 | 1152 | |||
XM_006713567.3 | XP_006713630.1 | 11 | 1257 | |||
XM_011512632.1 | XP_011510934.1 | 11 | 1205 | |||
XM_011512633.1 | XP_011510935.1 | 11 | 1259 | |||
XM_017006096.1 | XP_016861585.1 | 10 | 1152 | |||
XM_017006100.1 | XP_016861589.1 | 11 | 1257 | |||
XM_017006101.1 | XP_016861590.1 | 11 | 1167 | |||
GenBank mRNA | AF109184.1 | - | 12 | 1091 | ||
AF161541.1 | - | 12 | 1056 | |||
AK001721.1 | - | 10 | 900 | |||
AK001882.1 | - | 3 | 473 | |||
AK098500.1 | - | 2 | 1627 | |||
AK289577.1 | - | 12 | 1034 | |||
AK296837.1 | - | 10 | 869 | |||
AK299229.1 | - | 6 | 530 | |||
AK299769.1 | - | 9 | 812 | |||
AK304558.1 | - | 10 | 845 | |||
AK315932.1 | - | 6 | 535 | |||
BC013424.1 | - | 12 | 1092 | 3892143 | ||
BC032661.1 | - | 12 | 1202 | 5530425 | ||
BC041699.1 | - | 12 | 1017 | 5240423 |