Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015397.3 | NP_056212.1 | 9 | 1628 | ||
XM_011517843.1 | XP_011516145.1 | 9 | 1455 | |||
GenBank mRNA | AB067479.1 | - | 9 | 1548 | ||
AF131771.1 | - | 4 | 488 | |||
AK000354.1 | - | 9 | 1406 | |||
AK292935.1 | - | 9 | 1434 | |||
AL117646.1 | - | 6 | 762 | |||
BC008893.2 | - | 8 | 1191 | |||
BC063823.1 | - | 9 | 1546 | 6184796 | ||
DQ518909.1 | - | 9 | 1548 |