Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129727.2 | NP_001123199.1 | 11 | 1689 | ||
NM_001129728.1 | NP_001123200.1 | 10 | 1649 | |||
NM_001129729.2 | NP_001123201.1 | 10 | 1622 | |||
NM_001129731.2 | NP_001123203.1 | 8 | 1628 | |||
XM_011522985.2 | XP_011521287.1 | 10 | 1619 | |||
XM_011522986.2 | XP_011521288.1 | 10 | 1751 | |||
XM_011522987.2 | XP_011521289.1 | 11 | 1695 | |||
XM_011522988.2 | XP_011521290.1 | 10 | 1568 | |||
GenBank mRNA | AB197663.1 | - | 9 | 3885 | ||
AB197664.1 | - | 8 | 4052 | |||
AK024475.1 | - | 10 | 1584 | |||
AK304723.1 | - | 8 | 1040 | |||
AK308298.1 | - | 10 | 1558 | |||
AL117435.1 | - | 5 | 521 | |||
BC054486.1 | - | 8 | 1563 | 4820741 | ||
BC063501.1 | - | 10 | 1560 | 5527705 | ||
BC082974.1 | - | 11 | 1657 | 6291175 | ||
BC115390.1 | - | 7 | 1587 |