Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318520.1 | NP_001305449.1 | 33 | 4497 | ||
NM_173630.3 | NP_775901.3 | 34 | 4680 | |||
XM_006722434.3 | XP_006722497.1 | 34 | 4683 | |||
XM_006722435.3 | XP_006722498.1 | 34 | 4683 | |||
XM_011525902.2 | XP_011524204.1 | 33 | 4443 | |||
XM_011525903.2 | XP_011524205.1 | 32 | 4254 | |||
XM_011525904.2 | XP_011524206.1 | 34 | 4683 | |||
XM_011525905.2 | XP_011524207.1 | 34 | 4683 | |||
XM_011525906.2 | XP_011524208.1 | 24 | 3964 | |||
XM_011525907.2 | XP_011524209.1 | 34 | 4683 | |||
XM_017025693.1 | XP_016881182.1 | 33 | 4440 | |||
XM_017025694.1 | XP_016881183.1 | 31 | 4238 | |||
XM_017025695.1 | XP_016881184.1 | 27 | 3740 | |||
XM_017025696.1 | XP_016881185.1 | 34 | 4680 | |||
GenBank mRNA | AK126538.1 | - | 9 | 1125 | ||
AK128137.1 | - | 13 | 1833 | |||
BC047602.1 | - | 26 | 3588 | |||
BC143723.1 | - | 33 | 4474 | |||
CR749799.1 | - | 28 | 3896 |