Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318520.1 | NP_001305449.1 | 38, 39 | 5202 | ||
NM_173630.3 | NP_775901.3 | 39, 40 | 5385 | |||
XM_006722434.3 | XP_006722497.1 | 39, 40 | 5388 | |||
XM_006722435.3 | XP_006722498.1 | 39, 40 | 5388 | |||
XM_011525902.2 | XP_011524204.1 | 38, 39 | 5148 | |||
XM_011525903.2 | XP_011524205.1 | 37, 38 | 4959 | |||
XM_011525904.2 | XP_011524206.1 | 39, 40 | 5388 | |||
XM_011525906.2 | XP_011524208.1 | 29, 30 | 4669 | |||
XM_017025693.1 | XP_016881182.1 | 38, 39 | 5145 | |||
XM_017025694.1 | XP_016881183.1 | 36, 37 | 4943 | |||
XM_017025695.1 | XP_016881184.1 | 32, 33 | 4445 | |||
XM_017025696.1 | XP_016881185.1 | 39, 40 | 5385 | |||
GenBank mRNA | AK126538.1 | - | 14, 15 | 1830 | ||
AK128137.1 | - | 20, 21 | 2756 | |||
BC046222.2 | - | 4, 5 | 583 | |||
BC047602.1 | - | 31, 32 | 4293 | |||
BC143723.1 | - | 38, 39 | 5179 | |||
CR749799.1 | - | 33, 34 | 4601 |