Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015456.4 | NP_056271.3 | 9, 10 | 1442 | ||
GenBank mRNA | AB033008.1 | - | 1, 2 | 124 | ||
AF464935.1 | - | 9, 10 | 1247 | |||
AK022651.1 | - | 7, 8 | 938 | |||
AK091056.1 | - | 9, 10 | 1435 | |||
AL050280.1 | - | 6, 7 | 689 | |||
BC011892.1 | - | 8, 9 | 1151 | 3941719 | ||
BC019029.1 | - | 9, 10 | 1401 |