Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015457.2 | NP_056272.2 | 11 | 3161 | ||
XM_011544899.1 | XP_011543201.1 | 11 | 3091 | |||
XM_011544900.1 | XP_011543202.1 | 11 | 2999 | |||
XM_011544901.1 | XP_011543203.1 | 11 | 3122 | |||
XM_017017498.1 | XP_016872987.1 | 10 | 3205 | |||
GenBank mRNA | AB051535.1 | - | 11 | 3153 | ||
AK023130.1 | - | 10 | 2318 | |||
AK172807.1 | - | 7 | 1516 | |||
AL117662.1 | - | 1 | 384 | |||
AY894889.1 | - | 11 | 3267 | |||
BC026967.1 | - | 10 | 1904 | 3913317 | ||
BX647707.1 | - | 9 | 2153 |