Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303272.1 | NP_001290201.1 | 2 | 350 | ||
NM_015462.4 | NP_056277.2 | 2 | 350 | |||
GenBank mRNA | AK023702.1 | - | 2 | 240 | ||
AK299483.1 | - | 2 | 240 | |||
AY598333.1 | - | 2 | 235 | |||
BC064404.1 | - | 2 | 241 | 5555907 | ||
DA772356.1 | - | 2 | 350 |