Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005188.3 | NP_005179.2 | 16 | 2639 | ||
XM_017018511.1 | XP_016874000.1 | 15 | 2074 | |||
GenBank mRNA | BC132733.1 | - | 16 | 2568 | 40146755 | |
BC136463.1 | - | 16 | 2568 | 9020452 | ||
X57110.1 | - | 16 | 2645 |