Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013943.2 | NP_039234.1 | 5 | 840 | ||
GenBank mRNA | AF097330.1 | - | 5 | 695 | ||
AF109196.1 | - | 5 | 633 | |||
AK001624.1 | - | 5 | 727 | |||
AK095959.1 | - | 5 | 661 | |||
AK301466.1 | - | 5 | 638 | |||
AK307142.1 | - | 3 | 405 | |||
AK315051.1 | - | 5 | 677 | |||
AL117424.1 | - | 5 | 752 | |||
BC012444.1 | - | 5 | 705 | 3861372 | ||
CR533501.1 | - | 5 | 555 |