Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168278.1 | NP_001161750.1 | 7, 8 | 1276 | ||
NM_001168280.1 | NP_001161752.1 | 6, 7 | 1223 | |||
NM_015472.4 | NP_056287.1 | 6, 7 | 1359 | |||
XM_017006121.1 | XP_016861610.1 | 9, 10 | 1469 | |||
XM_017006122.1 | XP_016861611.1 | 7, 8 | 1268 | |||
GenBank mRNA | AJ299431.1 | - | 6, 7 | 1194 | ||
AK022036.1 | - | 7, 8 | 1276 | |||
AL050107.1 | - | 4, 5 | 583 | |||
AL833852.1 | - | 5, 6 | 1013 | |||
BC014052.2 | - | 6, 7 | 1253 | 3635491 |