Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004352.3 | NP_004343.1 | 1 | 616 | ||
GenBank mRNA | AK314276.1 | - | 1 | 590 | ||
BC093692.1 | - | 2 | 294 | 7939537 | ||
BC093718.1 | - | 2 | 294 | 7939563 | ||
M58583.1 | - | 1 | 564 |