Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIGD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099668.1 | NP_001093138.1 | 3, 4 | 519 | ||
NM_001099669.1 | NP_001093139.1 | 3, 4 | 372 | |||
NM_014056.3 | NP_054775.2 | 3, 4 | 348 | |||
GenBank mRNA | AF077034.1 | - | 3, 4 | 300 | ||
AF145385.1 | - | 3, 4 | 287 | |||
AK225461.1 | - | 3, 4 | 272 | |||
AK225571.1 | - | 3, 4 | 303 | |||
AK311815.1 | - | 3, 4 | 303 | |||
AL110233.1 | - | 3, 4 | 318 | |||
BC000601.1 | - | 3, 4 | 286 | 3346355 | ||
BC009583.1 | - | 3, 4 | 279 | 4093421 | ||
BC009594.1 | - | 3, 4 | 285 | 4109190 | ||
BC070277.1 | - | 3, 4 | 301 | 4686431 | ||
BG708126.1 | - | 3, 4 | 353 | |||
BP208784.1 | - | 3, 4 | 518 | |||
BP370118.1 | - | 3, 4 | 294 | |||
CR457385.1 | - | 2, 3 | 226 |